Tropylium tetrafluoroborate 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD156020-5GEA 1540 DKK
BLDPBD156020-5G BLDPBD156020-250MG BLDPBD156020-1G BLDPBD156020-25G
Tropylium tetrafluoroborate 98%
Tropylium tetrafluoroborate

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₇BF₄
Molekylvægt: 177,94 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00013402
CAS-nummer: 27081-10-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR