Ethyl-1-benzylpyrrolidine-3-carboxylate 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD15557-25GEA 985 DKK
BLDPBD15557-25G BLDPBD15557-5G BLDPBD15557-10G BLDPBD15557-1G
Ethyl-1-benzylpyrrolidine-3-carboxylate 97%
Ethyl-1-benzylpyrrolidine-3-carboxylate

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₄H₁₉NO₂
Molekylvægt: 233,31 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD04039852
CAS-nummer: 5747-92-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR