Tetrahydropyran-2-methanol 98%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD153503-1GEA
171
DKK
BLDPBD153503-1G
BLDPBD153503-25G
BLDPBD153503-5G
BLDPBD153503-250MG
Tetrahydropyran-2-methanol 98%
Tetrahydropyran-2-methanol
Storage condition: Sealed in dry, Room Temperature
Formel:
C₆H₁₂O₂ Molekylvægt: 116,16 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00006624 CAS-nummer: 100-72-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...