4-Acetyldiphenyl sulphide 97%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD15329-25GEA
1540
DKK
BLDPBD15329-25G
BLDPBD15329-1G
BLDPBD15329-5G
4-Acetyldiphenyl sulphide 97%
4-Acetyldiphenyl sulphide
Storage condition: Inert atmosphere, Room Temperature
Formel:
C₁₄H₁₂OS Molekylvægt: 228,32 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00026227 CAS-nummer: 10169-55-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...