Tetra-2-pyridinylpyrazine 97+%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD153200-250MGEA 512 DKK
BLDPBD153200-250MG BLDPBD153200-100MG
Tetra-2-pyridinylpyrazine 97+%
Tetra-2-pyridinylpyrazine

Storage condition: Inert atmosphere, Room Temperature

Formel: C₂₄H₁₆N₆
Molekylvægt: 388,42 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00014608
CAS-nummer: 25005-97-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR