3-Amino-6,6-dimethyl-1,4,5,6-tetrahydropyrrolo[3,4-c]pyrazole, N5-BOC protected 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD152828-1GEA 295 DKK
BLDPBD152828-1G BLDPBD152828-250MG BLDPBD152828-5G
3-Amino-6,6-dimethyl-1,4,5,6-tetrahydropyrrolo[3,4-c]pyrazole, N5-BOC protected 95%
3-Amino-6,6-dimethyl-1,4,5,6-tetrahydropyrrolo[3,4-c]pyrazole, N5-BOC protected

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₁₂H₂₀N₄O₂
Molekylvægt: 252,313 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD08275051
CAS-nummer: 398491-61-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR