Salicylaldehyde azine 97%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Salicylaldazine

BLDPBD15267-500GEA 3360 DKK
BLDPBD15267-500G BLDPBD15267-5G BLDPBD15267-100G BLDPBD15267-25G
Salicylaldehyde azine 97%
Salicylaldehyde azine

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₄H₁₂N₂O₂
Molekylvægt: 240,26 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00043496
CAS-nummer: 959-36-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR