2-Stearamidoacetic acid 98+%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD151931-1GEA 249 DKK
BLDPBD151931-1G BLDPBD151931-250MG BLDPBD151931-5G
2-Stearamidoacetic acid 98+%
2-Stearamidoacetic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₂₀H₃₉NO₃
Molekylvægt: 341,52 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00153472
CAS-nummer: 6333-54-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR