Sodium (S)-4-carboxy-2-dodecanamidobutanoate 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD151487-100GEA 311 DKK
BLDPBD151487-100G BLDPBD151487-25G
Sodium (S)-4-carboxy-2-dodecanamidobutanoate 95%
Sodium (S)-4-carboxy-2-dodecanamidobutanoate

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₇H₃₀NNaO₅
Molekylvægt: 351,4136 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01725209
CAS-nummer: 29923-31-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR