Sodium 4-(butoxycarbonyl)phenolate 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD151394-25GEA 62.1 DKK
BLDPBD151394-25G
Sodium 4-(butoxycarbonyl)phenolate 98%
Sodium 4-(butoxycarbonyl)phenolate

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₁H₁₃NaO₃
Molekylvægt: 216,2100 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01661516
CAS-nummer: 36457-20-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR