Rhodium(II) acetate dimer 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD150376-250MGEA 1620 DKK
BLDPBD150376-250MG BLDPBD150376-50MG BLDPBD150376-100MG BLDPBD150376-1G
Rhodium(II) acetate dimer 98%
Rhodium(II) acetate dimer

Storage condition: Inert atmosphere, Room Temperature

Formel: C₈H₁₂O₈Rh₂
Molekylvægt: 441,987 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003538
CAS-nummer: 15956-28-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR