Methoxatin Disodium 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD149951-250MGEA 5050 DKK
BLDPBD149951-250MG BLDPBD149951-1G
Methoxatin Disodium 98%
Methoxatin Disodium

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₄H₄N₂Na₂O₈
Molekylvægt: 374,16 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00151711
CAS-nummer: 122628-50-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR