3-(1-(tert-Butoxycarbonyl)piperidin-3-yl)propanoic acid 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD14985-1GEA 1060 DKK
BLDPBD14985-1G BLDPBD14985-250MG BLDPBD14985-100MG BLDPBD14985-5G
3-(1-(tert-Butoxycarbonyl)piperidin-3-yl)propanoic acid 95%
3-(1-(tert-Butoxycarbonyl)piperidin-3-yl)propanoic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₃H₂₃NO₄
Molekylvægt: 257,3260 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD02179008
CAS-nummer: 352004-58-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR