4-Methoxy-2-methylpyrimidine 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD149843-250MGEA 473 DKK
BLDPBD149843-250MG BLDPBD149843-100MG
4-Methoxy-2-methylpyrimidine 97%
4-Methoxy-2-methylpyrimidine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₆H₈N₂O
Molekylvægt: 124,14 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11111148
CAS-nummer: 7314-65-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR