4-Pyridineacetamide 95%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD149763-250MGEA
287
DKK
BLDPBD149763-250MG
BLDPBD149763-1G
BLDPBD149763-5G
4-Pyridineacetamide 95%
4-Pyridineacetamide
Storage condition: Inert atmosphere, Room Temperature
Formel:
C₇H₈N₂O Molekylvægt: 136,15 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD02685124 CAS-nummer: 39640-62-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...