Pyrazolo[1,5-a]pyridin-2-ylmethanol 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD149625-1GEA 3310 DKK
BLDPBD149625-1G BLDPBD149625-100MG BLDPBD149625-250MG
Pyrazolo[1,5-a]pyridin-2-ylmethanol 95%
Pyrazolo[1,5-a]pyridin-2-ylmethanol

Storage condition: Sealed in dry, 2-8°C

Formel: C₈H₈N₂O
Molekylvægt: 148,16 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD09878923
CAS-nummer: 76943-47-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR