Potassiummyristate 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD148840-5GEA 125 DKK
BLDPBD148840-5G BLDPBD148840-100G BLDPBD148840-25G
Potassiummyristate 95%
Potassiummyristate

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₄H₂₇KO₂
Molekylvægt: 266,46 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00152765
CAS-nummer: 13429-27-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR