(1,3-Dioxolan-2-ylmethyl)triphenylphosphonium bromide 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD1480-10GEA 77.6 DKK
BLDPBD1480-10G BLDPBD1480-25G BLDPBD1480-5G BLDPBD1480-500G BLDPBD1480-100G
(1,3-Dioxolan-2-ylmethyl)triphenylphosphonium bromide 97%
(1,3-Dioxolan-2-ylmethyl)triphenylphosphonium bromide

Storage condition: Inert atmosphere, Room Temperature

Formel: C₂₂H₂₂BrO₂P
Molekylvægt: 429,30 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011966
CAS-nummer: 52509-14-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR