2-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetic acid 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD147617-1GEA 628 DKK
BLDPBD147617-1G BLDPBD147617-5G BLDPBD147617-250MG
2-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetic acid 98%
2-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetic acid

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₄H₁₉BO₄
Molekylvægt: 262,1100 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD04973389
CAS-nummer: 797755-05-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR