Phenoxyacetic anhydride 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD147506-1GEA 62.1 DKK
BLDPBD147506-1G BLDPBD147506-25G BLDPBD147506-0.025 BLDPBD147506-5G
Phenoxyacetic anhydride 98%
Phenoxyacetic anhydride

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₆H₁₄O₅
Molekylvægt: 286,2794 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00059681
CAS-nummer: 14316-61-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR