Phenolphthalin 95%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD147483-5GEA 163 DKK
BLDPBD147483-5G BLDPBD147483-0.025 BLDPBD147483-1G BLDPBD147483-25G
Phenolphthalin 95%
Phenolphthalin

Storage condition: Sealed in dry, Room Temperature

Formel: C₂₀H₁₆O₄
Molekylvægt: 320,3387 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00036135
CAS-nummer: 81-90-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR