Phenethylbiguanide hydrochloride 98%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD147439-1GEA 46.6 DKK
BLDPBD147439-1G BLDPBD147439-5G BLDPBD147439-10G BLDPBD147439-100G BLDPBD147439-25G
Phenethylbiguanide hydrochloride 98%
Phenethylbiguanide hydrochloride

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₁₀H₁₆ClN₅
Molekylvægt: 241,7205 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00035039
CAS-nummer: 834-28-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR