1-Ethyl-6-fluoro-7-(4-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid monomethanesulphonate 98+%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD146879-10GEA 559 DKK
BLDPBD146879-10G BLDPBD146879-5G BLDPBD146879-1G
1-Ethyl-6-fluoro-7-(4-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid monomethanesulphonate 98+%
1-Ethyl-6-fluoro-7-(4-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid monomethanesulphonate

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₁₈H₂₄FN₃O₆S
Molekylvægt: 429,46 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD09028102
CAS-nummer: 70458-95-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR