Palmitic anhydride 96%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD146673-25GEA
745
DKK
BLDPBD146673-25G
BLDPBD146673-0.025
BLDPBD146673-5G
Palmitic anhydride 96%
Palmitic anhydride
Storage condition: Inert atmosphere, Room Temperature
Formel:
C₃₂H₆₂O₃ Molekylvægt: 494,8329 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00008992 CAS-nummer: 623-65-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...