2-(Benzyloxy)-4-nitroaniline 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD14626-1GEA 4410 DKK
BLDPBD14626-1G BLDPBD14626-100MG BLDPBD14626-250MG
2-(Benzyloxy)-4-nitroaniline 95%
2-(Benzyloxy)-4-nitroaniline

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₁₃H₁₂N₂O₃
Molekylvægt: 244,24 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00276800
CAS-nummer: 25945-96-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR