2-Benzyloxy-5-fluorophenylboronic acid 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD14609-5GEA 605 DKK
BLDPBD14609-5G BLDPBD14609-25G BLDPBD14609-1G BLDPBD14609-10G
2-Benzyloxy-5-fluorophenylboronic acid 98%
2-Benzyloxy-5-fluorophenylboronic acid

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₃H₁₂BFO₃
Molekylvægt: 246,0420 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03788403
CAS-nummer: 779331-47-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR