2-Methoxydibenzo[b,d]furan-3-amine 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD14588-5GEA 264 DKK
BLDPBD14588-5G BLDPBD14588-25G BLDPBD14588-1G
2-Methoxydibenzo[b,d]furan-3-amine 98%
2-Methoxydibenzo[b,d]furan-3-amine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₃H₁₁NO₂
Molekylvægt: 213,23 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011554
CAS-nummer: 5834-17-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR