Ethyl-4-hydroxy-6-(trifluoromethoxy)quinoline-3-carboxylate 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD14513-5GEA 287 DKK
BLDPBD14513-5G BLDPBD14513-10G BLDPBD14513-1G BLDPBD14513-25G
Ethyl-4-hydroxy-6-(trifluoromethoxy)quinoline-3-carboxylate 95%
Ethyl-4-hydroxy-6-(trifluoromethoxy)quinoline-3-carboxylate

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₃H₁₀F₃NO₄
Molekylvægt: 301,2200 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00219847
CAS-nummer: 175203-85-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR