3-Pyrid-3-ylbenzoic acid 97%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD14468-5GEA
1780
DKK
BLDPBD14468-5G
BLDPBD14468-250MG
BLDPBD14468-1G
3-Pyrid-3-ylbenzoic acid 97%
3-Pyrid-3-ylbenzoic acid
Storage condition: Sealed in dry, Room Temperature
Formel:
C₁₂H₉NO₂ Molekylvægt: 199,2054 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD03426512 CAS-nummer: 4385-77-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...