N-Ethylacrylamide 98% stabilised

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

BLDPBD144219-5GEA 2650 DKK
BLDPBD144219-5G BLDPBD144219-250MG BLDPBD144219-100MG BLDPBD144219-25G BLDPBD144219-1G
N-Ethylacrylamide 98% stabilised
N-Ethylacrylamide

Storage condition: Sealed in dry, 2-8°C

Formel: C₅H₉NO
Molekylvægt: 99,1311 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00048514
CAS-nummer: 5883-17-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR