3-Butylidenephthalide mixture of isomers 97%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD143786-100GEA
807
DKK
BLDPBD143786-100G
BLDPBD143786-5G
BLDPBD143786-25G
3-Butylidenephthalide mixture of isomers 97%
3-Butylidenephthalide
Storage condition: Keep in dark place, Sealed in dry, 2-8°C
Formel:
C₁₂H₁₂O₂ Molekylvægt: 188,23 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00047319 CAS-nummer: 551-08-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...