1-Bromo-3-(4-fluorophenoxy)benzene 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD14375-1GEA 838 DKK
BLDPBD14375-1G BLDPBD14375-5G BLDPBD14375-250MG
1-Bromo-3-(4-fluorophenoxy)benzene 97%
1-Bromo-3-(4-fluorophenoxy)benzene

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₂H₈BrFO
Molekylvægt: 267,0937 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00142589
CAS-nummer: 50904-38-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR