N-Tosylbenzamide 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD143388-250MGEA 326 DKK
BLDPBD143388-250MG BLDPBD143388-1G BLDPBD143388-100MG BLDPBD143388-5G BLDPBD143388-25G
N-Tosylbenzamide 95%
N-Tosylbenzamide

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₄H₁₃NO₃S
Molekylvægt: 275,3229 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00431862
CAS-nummer: 6971-74-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR