9-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-2-(methylamino)-1H-purin-6(9H)-one 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD142795-25MGEA 2910 DKK
BLDPBD142795-25MG BLDPBD142795-10MG BLDPBD142795-50MG BLDPBD142795-100MG
9-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-2-(methylamino)-1H-purin-6(9H)-one 95%
9-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-2-(methylamino)-1H-purin-6(9H)-one

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₁₁H₁₅N₅O₅
Molekylvægt: 297,26 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00057039
CAS-nummer: 2140-77-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR