4-(Methylamino)phenol sulphate 98%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: MetolBis(4-hydroxy-N-methylanilinium)sulfat

BLDPBD140041-10GEA 69.8 DKK
BLDPBD140041-10G BLDPBD140041-100G BLDPBD140041-25G
4-(Methylamino)phenol sulphate 98%
Bis(4-hydroxy-N-methylanilinium)sulfat

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₄H₂₀N₂O₆S
Molekylvægt: 344,39 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00058280
CAS-nummer: 51-72-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR