4-[4-(Trifluoromethoxy)phenoxy]piperidine 97%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD13966-1GEA 194 DKK
BLDPBD13966-1G BLDPBD13966-100G BLDPBD13966-10G BLDPBD13966-250MG BLDPBD13966-5G BLDPBD13966-25G
4-[4-(Trifluoromethoxy)phenoxy]piperidine 97%
4-[4-(Trifluoromethoxy)phenoxy]piperidine

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₁₂H₁₄F₃NO₂
Molekylvægt: 261,25 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD06656166
CAS-nummer: 287952-67-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR