Methyl imidazo[1,2-a]pyridine-6-carboxylate 98%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD139461-250MGEA 132 DKK
BLDPBD139461-250MG BLDPBD139461-1G BLDPBD139461-5G
Methyl imidazo[1,2-a]pyridine-6-carboxylate 98%
Methyl imidazo[1,2-a]pyridine-6-carboxylate

Storage condition: Sealed in dry, 2-8°C

Formel: C₉H₈N₂O₂
Molekylvægt: 176,1720 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD06200984
CAS-nummer: 136117-69-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR