6-Acetamido-1,2,3,4-tetrahydronaphthalen-1-one 96%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD13942-250MGEA 256 DKK
BLDPBD13942-250MG BLDPBD13942-1G
6-Acetamido-1,2,3,4-tetrahydronaphthalen-1-one 96%
6-Acetamido-1,2,3,4-tetrahydronaphthalen-1-one

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₂H₁₃NO₂
Molekylvægt: 203,237 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00099461
CAS-nummer: 88611-67-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR