3-Methoxy-5-(methoxycarbonyl)benzoic acid 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD139174-5GEA 2970 DKK
BLDPBD139174-5G BLDPBD139174-250MG BLDPBD139174-100MG BLDPBD139174-1G
3-Methoxy-5-(methoxycarbonyl)benzoic acid 95%
3-Methoxy-5-(methoxycarbonyl)benzoic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₁₀O₅
Molekylvægt: 210,18 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03840746
CAS-nummer: 71590-08-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR