Methyl but-3-enoate 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD138623-250MGEA 117 DKK
BLDPBD138623-250MG BLDPBD138623-5G BLDPBD138623-1G
Methyl but-3-enoate 97%
Methyl but-3-enoate

Storage condition: Sealed in dry, Store in freezer, under -20°C

Formel: C₅H₈O₂
Molekylvægt: 100,1158 g/mol
Opbevaringstemperatur: Fryser
MDL Number: MFCD00082587
CAS-nummer: 3724-55-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR