Methyl-2,3-diaminobenzoate 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD138064-5GEA 1190 DKK
BLDPBD138064-5G BLDPBD138064-250MG BLDPBD138064-100MG BLDPBD138064-25G BLDPBD138064-1G
Methyl-2,3-diaminobenzoate 97%
Methyl-2,3-diaminobenzoate

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₈H₁₀N₂O₂
Molekylvægt: 166,1800 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD04038589
CAS-nummer: 107582-20-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR