Methyl10-oxooctadecanoate 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD137896-1GEA 18970 DKK
BLDPBD137896-1G BLDPBD137896-100MG BLDPBD137896-50MG BLDPBD137896-250MG
Methyl10-oxooctadecanoate 95%
Methyl10-oxooctadecanoate

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₉H₃₆O₃
Molekylvægt: 312,48 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD05864164
CAS-nummer: 1842-70-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR