1-Chloro-3-propylbenzene 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD137271-250MGEA 1490 DKK
BLDPBD137271-250MG BLDPBD137271-1G BLDPBD137271-100MG
1-Chloro-3-propylbenzene 95%
1-Chloro-3-propylbenzene

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₁₁Cl
Molekylvægt: 154,63 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09033164
CAS-nummer: 57430-24-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR