Magnesiumchlorid 97%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD136984-100GEA 54.3 DKK
BLDPBD136984-100G BLDPBD136984-500G BLDPBD136984-25G BLDPBD136984-1000G BLDPBD136984-0.5G
Magnesiumchlorid 97%
Magnesium chloride

Storage condition: Inert atmosphere, Room Temperature

Formel: MgCl₂
Molekylvægt: 95,21 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011106
CAS-nummer: 7786-30-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR