Sodium (S)-5-oxopyrrolidine-2-carboxylate 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD136718-100GEA 202 DKK
BLDPBD136718-100G BLDPBD136718-1000G BLDPBD136718-25G BLDPBD136718-1G BLDPBD136718-500G
Sodium (S)-5-oxopyrrolidine-2-carboxylate 97%
Sodium (S)-5-oxopyrrolidine-2-carboxylate

Storage condition: Inert atmosphere, Room Temperature

Formel: C₅H₆NNaO₃
Molekylvægt: 151,09 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01749005
CAS-nummer: 28874-51-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR