L-Cystine dihydrochloride 98%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD135630-25GEA
148
DKK
BLDPBD135630-25G
BLDPBD135630-5G
L-Cystine dihydrochloride 98%
L-Cystine dihydrochloride
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formel:
C₆H₁₂N₂O₄S₂·2HCl Molekylvægt: 313,23 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00070399 CAS-nummer: 30925-07-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...