16-Methylheptadecanoic acid 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD135403-25GEA 46.6 DKK
BLDPBD135403-25G BLDPBD135403-100G BLDPBD135403-500G
16-Methylheptadecanoic acid 97%
16-Methylheptadecanoic acid

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₈H₃₆O₂
Molekylvægt: 284,47 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00044082
CAS-nummer: 2724-58-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR