Isopropylthioglycolate 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD135298-5GEA 2920 DKK
BLDPBD135298-5G BLDPBD135298-250MG BLDPBD135298-1G BLDPBD135298-100MG
Isopropylthioglycolate 95%
Isopropylthioglycolate

Storage condition: Inert atmosphere, Room Temperature

Formel: C₅H₁₀O₂S
Molekylvægt: 134,19 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00022078
CAS-nummer: 7383-61-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR