Iridium (III) chlorid hydrat 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD134883-1GEA 2160 DKK
BLDPBD134883-1G BLDPBD134883-100MG BLDPBD134883-5G BLDPBD134883-250MG
Iridium (III) chlorid hydrat 98%
Iridium (III) chlorid hydrat

Storage condition: Inert atmosphere, Room Temperature

Formel: Cl₃Ir·xH₂O
Molekylvægt: 298,58 g/mol (anhy)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00149749
CAS-nummer: 14996-61-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR