N-Methyl-3,4,5-trimethoxybenzylamine 95%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD13426-1GEA
1050
DKK
BLDPBD13426-1G
BLDPBD13426-250MG
N-Methyl-3,4,5-trimethoxybenzylamine 95%
N-Methyl-3,4,5-trimethoxybenzylamine
Storage condition: Keep in dark place, Inert atmosphere, 2-8°C
Formel:
C₁₁H₁₇NO₃ Molekylvægt: 211,26 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00798597 CAS-nummer: 58780-82-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...